Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302489.1 | NP_001289418.1 | 4, 5 | 477 | ||
NM_001302490.1 | NP_001289419.1 | 3, 4 | 409 | |||
NM_001610.3 | NP_001601.1 | 4, 5 | 505 | |||
GenBank mRNA | AK298439.1 | - | 4, 5 | 480 | ||
AK300569.1 | - | 3, 4 | 433 | |||
AK301860.1 | - | 3, 4 | 367 | |||
AK308512.1 | - | 2, 3 | 724 | |||
AY007137.1 | - | 1, 2 | 37 | |||
BC003160.1 | - | 4, 5 | 450 | 2957907 | ||
BC093010.1 | - | 4, 5 | 463 | 30332133 | ||
X12548.1 | - | 4, 5 | 459 |