Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTL6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004301.4 | NP_004292.1 | 11, 12 | 1243 | ||
NM_177989.3 | NP_817126.1 | 11, 12 | 1099 | |||
NM_178042.3 | NP_829888.1 | 11, 12 | 1291 | |||
GenBank mRNA | AB015907.1 | - | 11, 12 | 1136 | ||
AB060168.1 | - | 11, 12 | 1068 | |||
AB061315.1 | - | 11, 12 | 1213 | |||
AF041474.1 | - | 11, 12 | 1166 | |||
AK021756.1 | - | 11, 12 | 1159 | |||
AK098691.1 | - | 11, 12 | 1197 | |||
AK223212.1 | - | 11, 12 | 1164 | |||
AL136608.1 | - | 11, 12 | 1183 | |||
BC000949.2 | - | 11, 12 | 1041 | 3445923 | ||
BC001391.2 | - | 11, 12 | 1084 | 3050818 | ||
BC036371.1 | - | 11, 12 | 1182 | 4827785 | ||
CR533529.1 | - | 11, 12 | 1030 |