Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACVR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105.4 | NP_001096.1 | 9 | 1608 | ||
NM_001111067.2 | NP_001104537.1 | 9 | 1427 | |||
XM_005246939.3 | XP_005246996.1 | 9 | 1479 | |||
XM_005246940.3 | XP_005246997.1 | 9 | 1447 | |||
XM_006712825.3 | XP_006712888.1 | 9 | 1560 | |||
XM_011512106.2 | XP_011510408.1 | 10 | 1527 | |||
XM_011512107.2 | XP_011510409.1 | 9 | 1499 | |||
XM_011512108.2 | XP_011510410.1 | 9 | 2906 | |||
GenBank mRNA | AK313630.1 | - | 9 | 1403 | ||
BC033867.1 | - | 9 | 1608 | 5209711 | ||
BC036748.1 | - | 9 | 1608 | |||
KU177885.1 | - | 7 | 1178 | |||
L02911.1 | - | 10 | 1518 | |||
Z22534.1 | - | 8 | 1281 |