Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000666.2 | NP_000657.1 | 12, 13 | 1233 | ||
NM_001198895.1 | NP_001185824.1 | 12, 13 | 1228 | |||
NM_001198896.1 | NP_001185825.1 | 10, 11 | 1017 | |||
NM_001198897.1 | NP_001185826.1 | 10, 11 | 1038 | |||
NM_001198898.1 | NP_001185827.1 | 11, 12 | 1128 | |||
GenBank mRNA | BC000545.2 | - | 12, 13 | 977 | 3162222 | |
BC003023.1 | - | 12, 13 | 970 | 2821665 | ||
BC014112.2 | - | 12, 13 | 953 | 4543442 | ||
CD014023.1 | - | 10, 11 | 858 | |||
CD014026.1 | - | 10, 11 | 879 | |||
CD014027.1 | - | 11, 12 | 969 | |||
D14524.1 | - | 12, 13 | 967 | |||
D16307.1 | - | 12, 13 | 965 | |||
EU794586.1 | - | 12, 13 | 953 | |||
KU177890.1 | - | 11, 12 | 909 | |||
L07548.1 | - | 12, 13 | 970 |
Gene Symbol: ABHD14A-ACY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316331.1 | NP_001303260.1 | 14, 15 | 1434 | ||
GenBank mRNA | AK298086.1 | - | 14, 15 | 1410 |