Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACYP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107.4 | NP_001098.1 | 3 | 320 | ||
NM_001302616.1 | NP_001289545.1 | 3 | 302 | |||
NM_001302617.1 | NP_001289546.1 | 3 | 498 | |||
NR_126393.1 | 4 | 399 | ||||
NR_126394.1 | 2 | 406 | ||||
NR_126395.1 | 2 | 228 | ||||
GenBank mRNA | AK312101.1 | - | 3 | 320 | ||
BC035568.1 | - | 3 | 305 | 5392248 | ||
BE467248.1 | - | 1 | 314 | |||
BG614847.1 | - | 4 | 366 | |||
BP316920.1 | - | 3 | 283 | |||
BX104912.1 | - | 2 | 173 | |||
CD675334.1 | - | 3 | 507 | |||
DB071115.1 | - | 3 | 320 | |||
HY012032.1 | - | 2 | 368 | |||
X84194.1 | - | 2 | 284 |