Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025107.2 | NP_001020278.1 | 10 | 2917 | ||
NM_001111.4 | NP_001102.2 | 10 | 3048 | |||
NM_001193495.1 | NP_001180424.1 | 10 | 3086 | |||
NM_015840.3 | NP_056655.2 | 10 | 2970 | |||
NM_015841.3 | NP_056656.2 | 10 | 2913 | |||
XM_006711109.2 | XP_006711172.1 | 10 | 2854 | |||
XM_006711111.3 | XP_006711174.1 | 11 | 2976 | |||
XM_006711112.2 | XP_006711175.1 | 11 | 2678 | |||
XM_006711113.2 | XP_006711176.1 | 11 | 2752 | |||
XM_011509060.1 | XP_011507362.1 | 10 | 2950 | |||
XM_011509061.2 | XP_011507363.2 | 10 | 2839 | |||
XM_011509062.1 | XP_011507364.1 | 10 | 2877 | |||
XM_017000037.1 | XP_016855526.1 | 11 | 2767 | |||
GenBank mRNA | AB209891.1 | - | 10 | 2861 | ||
AK304153.1 | - | 10 | 3002 | |||
BC038227.1 | - | 10 | 2975 | 5497548 | ||
BE886208.1 | - | 338 | ||||
BX538232.1 | - | 10 | 2901 | |||
BX640741.1 | - | 10 | 2899 | |||
KJ544856.1 | - | 9 | 2806 | |||
U10439.1 | - | 10 | 2960 | |||
U18121.1 | - | 10 | 2853 | |||
X79448.1 | - | 10 | 3086 | |||
X79449.1 | - | 10 | 3008 | |||
X98559.1 | - | 9 | 2791 |