Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020546.2 | NP_065433.2 | 8 | 1195 | ||
XM_011513942.2 | XP_011512244.1 | 8 | 1142 | |||
GenBank mRNA | AB028983.1 | - | 5 | 516 | ||
AF410885.1 | - | 8 | 1154 | |||
AK123013.1 | - | 7 | 1096 | |||
AK127036.1 | - | 8 | 1143 | |||
AK294555.1 | - | 5 | 651 | |||
AK295531.1 | - | 5 | 535 | |||
AK299775.1 | - | 7 | 675 | |||
AK304041.1 | - | 8 | 1030 | |||
BC111743.1 | - | 8 | 1133 | 40038765 |