Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADH1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000669.4 | NP_000660.1 | 8, 9 | 1462 | ||
NR_133005.1 | 8, 9 | 1418 | ||||
XM_011531588.2 | XP_011529890.1 | 7, 8 | 1252 | |||
XM_011531589.1 | XP_011529891.1 | 9, 10 | 1582 | |||
GenBank mRNA | BC062476.1 | - | 8, 9 | 1158 | 30408761 | |
BC066227.1 | - | 8, 9 | 1120 | 7002195 | ||
BC066228.1 | - | 8, 9 | 1120 | 7002197 | ||
BC067419.1 | - | 8, 9 | 1120 | 7002191 | ||
BC067420.1 | - | 8, 9 | 1120 | 7002192 | ||
BC067421.1 | - | 8, 9 | 1120 | 7002193 | ||
BC067422.1 | - | 8, 9 | 1120 | 7002194 | ||
BC074771.2 | - | 8, 9 | 1120 | 30915457 | ||
BC074786.2 | - | 8, 9 | 1119 | 30915344 | ||
CD014078.1 | - | 8, 9 | 1119 | |||
M12272.1 | - | 8, 9 | 1172 | |||
X04299.1 | - | 8, 9 | 1164 | |||
X04350.1 | - | 6, 7 | 934 |