Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242837.1 | NP_001229766.1 | 14 | 2049 | ||
NM_012305.3 | NP_036437.1 | 14 | 2046 | |||
XM_011519929.1 | XP_011518231.1 | 14 | 1806 | |||
XM_011519930.2 | XP_011518232.1 | 14 | 2010 | |||
GenBank mRNA | AB020706.2 | - | 14 | 1833 | ||
AK027640.1 | - | 6 | 783 | |||
AK027891.1 | - | 14 | 1958 | |||
AK223558.1 | - | 14 | 1977 | |||
AK293763.1 | - | 9 | 1122 | |||
AK299364.1 | - | 14 | 1824 | |||
BC006155.1 | - | 14 | 1974 | 3627239 |