Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242837.1 | NP_001229766.1 | 7 | 938 | ||
NM_012305.3 | NP_036437.1 | 7 | 938 | |||
XM_011519929.1 | XP_011518231.1 | 7 | 695 | |||
XM_011519930.2 | XP_011518232.1 | 7 | 899 | |||
GenBank mRNA | AB020706.2 | - | 7 | 725 | ||
AB209422.1 | - | 4 | 3380 | |||
AK001986.1 | - | 7 | 867 | |||
AK027891.1 | - | 7 | 847 | |||
AK075497.1 | - | 4 | 417 | |||
AK223558.1 | - | 7 | 866 | |||
AK299364.1 | - | 7 | 716 | |||
BC006155.1 | - | 7 | 866 | 3627239 |