Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP2B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030006.1 | NP_001025177.1 | 21 | 2943 | ||
NM_001282.2 | NP_001273.1 | 20 | 2901 | |||
XM_005257937.3 | XP_005257994.1 | 21 | 3008 | |||
XM_005257938.2 | XP_005257995.1 | 21 | 2909 | |||
XM_011524448.1 | XP_011522750.1 | 22 | 3403 | |||
XM_011524449.2 | XP_011522751.1 | 22 | 3104 | |||
XM_011524450.1 | XP_011522752.1 | 22 | 3005 | |||
XM_011524451.1 | XP_011522753.1 | 21 | 2932 | |||
XM_011524452.1 | XP_011522754.1 | 23 | 3172 | |||
XM_011524453.1 | XP_011522755.1 | 23 | 3090 | |||
XM_011524454.1 | XP_011522756.1 | 23 | 3106 | |||
XM_011524455.2 | XP_011522757.1 | 21 | 2952 | |||
XM_017024284.1 | XP_016879773.1 | 20 | 2966 | |||
XM_017024285.1 | XP_016879774.1 | 22 | 3076 | |||
XM_017024286.1 | XP_016879775.1 | 21 | 2999 | |||
GenBank mRNA | AK289528.1 | - | 21 | 2884 | ||
AK292531.1 | - | 21 | 2821 | |||
AK295667.1 | - | 10 | 1093 | |||
AK301522.1 | - | 21 | 2778 | |||
AK304126.1 | - | 17 | 2298 | |||
AY341427.1 | - | 21 | 2936 | |||
BC006201.2 | - | 21 | 2850 | 3532472 | ||
BC012150.1 | - | 12 | 1571 | |||
CR749392.1 | - | 21 | 2926 | |||
M34175.1 | - | 20 | 2889 |