Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRISP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131.2 | NP_001122.2 | 2 | 130 | ||
NM_001205220.1 | NP_001192149.1 | 2 | 141 | |||
NM_170609.1 | NP_733758.1 | 2 | 130 | |||
GenBank mRNA | AB451334.1 | - | 1 | 51 | ||
AB451479.1 | - | 1 | 51 | |||
AK292497.1 | - | 2 | 111 | |||
BC131707.1 | - | 2 | 101 | |||
GQ472203.1 | - | 2 | 124 | |||
X95237.1 | - | 2 | 124 | |||
X95238.1 | - | 2 | 124 |