Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134.2 | NP_001125.1 | 2 | 159 | ||
GenBank mRNA | AF484525.1 | - | 3 | 296 | ||
AF484526.1 | - | 2 | 176 | |||
AK297669.1 | - | 2 | 157 | |||
AK297674.1 | - | 2 | 154 | |||
AK314817.1 | - | 2 | 153 | |||
BC027881.1 | - | 2 | 145 | 5208299 | ||
V01514.1 | - | 2 | 159 |