Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000027.3 | NP_000018.2 | 8 | 966 | ||
NM_001171988.1 | NP_001165459.1 | 8 | 936 | |||
NR_033655.1 | 7 | 890 | ||||
GenBank mRNA | AK312982.1 | - | 8 | 900 | ||
AW467768.1 | - | 1 | 99 | |||
BC012392.1 | - | 8 | 889 | 3859317 | ||
CR541715.1 | - | 8 | 838 | |||
M64073.1 | - | 8 | 841 | |||
X55762.1 | - | 8 | 865 |