Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000029.3 | NP_000020.1 | 4 | 1664 | ||
GenBank mRNA | AB209769.1 | - | 4 | 1176 | ||
AK222798.1 | - | 4 | 1195 | |||
AK222815.1 | - | 4 | 1196 | |||
AK293507.1 | - | 5 | 949 | |||
AK303755.1 | - | 5 | 1137 | |||
AK307978.1 | - | 5 | 1110 | |||
AK312291.1 | - | 4 | 1274 | |||
BC011519.1 | - | 4 | 1184 | 4213559 | ||
BT006851.1 | - | 3 | 1156 | |||
K02215.1 | - | 4 | 1195 | |||
T97863.1 | - | 2 | 53 | |||
W01494.1 | - | 2 | 139 |