Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APLNR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005161.4 | NP_005152.1 | 1 | 1103 | ||
NR_027991.1 | 1 | 1103 | ||||
GenBank mRNA | AK074514.1 | - | 2 | 545 | ||
AK075252.1 | - | 1 | 851 | |||
AK097232.1 | - | 2 | 490 | |||
AK315542.1 | - | 1 | 900 | |||
AL548962.3 | - | 1 | 835 | |||
BC032688.1 | - | 1 | 885 | 5241995 | ||
X89271.1 | - | 1 | 963 |
Set Membership: |
G-protein coupled receptor |