Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000477.5 | NP_000468.1 | 14 | 1887 | ||
GenBank mRNA | A06977.1 | - | 14 | 1889 | ||
AF190168.1 | - | 14 | 1814 | |||
AK292755.1 | - | 14 | 1852 | |||
AK298437.1 | - | 14 | 1880 | |||
AK298461.1 | - | 14 | 1597 | |||
AK314794.1 | - | 14 | 1855 | |||
AY544124.1 | - | 10 | 1245 | |||
AY550967.1 | - | 14 | 1821 | |||
AY728024.1 | - | 14 | 1820 | |||
AY960291.1 | - | 14 | 1846 | |||
BC014308.1 | - | 5 | 592 | |||
BC034023.1 | - | 14 | 1855 | 4734617 | ||
BC035969.1 | - | 10 | 1216 | 4714468 | ||
BC036003.1 | - | 14 | 1855 | 4724105 | ||
BC041789.1 | - | 10 | 1279 | 4766983 | ||
CR749331.1 | - | 14 | 1854 | |||
DQ986150.1 | - | 14 | 1825 | |||
HQ537426.1 | - | 13 | 1742 | |||
V00494.1 | - | 14 | 1853 | |||
V00495.1 | - | 14 | 1889 |