Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000690.3 | NP_000681.2 | 12 | 1544 | ||
NM_001204889.1 | NP_001191818.1 | 11 | 1403 | |||
GenBank mRNA | AK223373.1 | - | 12 | 1486 | ||
AK301375.1 | - | 11 | 1341 | |||
AK314856.1 | - | 12 | 1465 | |||
AY621070.1 | - | 12 | 1509 | |||
BC002967.1 | - | 12 | 1494 | 3543343 | ||
BC071839.1 | - | 12 | 1453 | 5477768 | ||
CB851683.1 | - | 2 | 529 | |||
CR456991.1 | - | 12 | 1448 | |||
K03001.1 | - | 9 | 1094 | |||
M26760.1 | - | 9 | 1094 | |||
M54931.1 | - | 7 | 809 | |||
X05409.1 | - | 12 | 1481 | |||
Y00109.1 | - | 12 | 1472 |