Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH3B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000694.3 | NP_000685.1 | 5 | 501 | ||
NM_001030010.2 | NP_001025181.1 | 4 | 390 | |||
NM_001161473.2 | NP_001154945.1 | 5 | 559 | |||
NM_001290058.1 | NP_001276987.1 | 5 | 501 | |||
NM_001290059.1 | NP_001276988.1 | 5 | 501 | |||
GenBank mRNA | AB209651.1 | - | 3 | 2656 | ||
AK291505.1 | - | 5 | 484 | |||
BC002553.2 | - | 5 | 483 | |||
BC013584.2 | - | 5 | 454 | 3878124 | ||
BC033099.1 | - | 4 | 328 | 5446816 | ||
BM561843.1 | - | 5 | 500 | |||
BT009832.1 | - | 4 | 412 | |||
CD014083.1 | - | 4 | 426 | |||
CD014084.1 | - | 4 | 426 | |||
DA624671.1 | - | 5 | 504 | |||
EF411198.1 | - | 5 | 528 | |||
U10868.1 | - | 5 | 459 |