Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005164.3 | NP_005155.1 | 3 | 1621 | ||
XM_011538027.2 | XP_011536329.1 | 4 | 1561 | |||
XM_017018992.1 | XP_016874481.1 | 3 | 1621 | |||
XM_017018993.1 | XP_016874482.1 | 3 | 1621 | |||
GenBank mRNA | AJ000327.1 | - | 3 | 1330 | ||
AK314254.1 | - | 3 | 1621 | |||
BC104901.1 | - | 3 | 1227 | 8143904 | ||
BC104903.1 | - | 3 | 1221 | 8143906 |