Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000034.3 | NP_000025.1 | 12 | 1805 | ||
NM_001127617.2 | NP_001121089.1 | 7 | 1027 | |||
NM_001243177.1 | NP_001230106.1 | 8 | 1148 | |||
NM_184041.2 | NP_908930.1 | 7 | 994 | |||
NM_184043.2 | NP_908932.1 | 7 | 966 | |||
XM_011545768.2 | XP_011544070.1 | 9 | 1668 | |||
GenBank mRNA | AK026577.1 | - | 7 | 843 | ||
AK098778.1 | - | 10 | 1846 | |||
AK301993.1 | - | 8 | 960 | |||
BC000367.2 | - | 4 | 406 | |||
BC004333.1 | - | 12 | 1725 | 3630974 | ||
BC010660.1 | - | 7 | 912 | 3869603 | ||
BC012880.1 | - | 7 | 840 | 3459009 | ||
BC013614.1 | - | 7 | 923 | 3880857 | ||
BC015888.2 | - | 7 | 836 | 3911623 | ||
BC016170.1 | - | 4 | 406 | |||
BC016800.1 | - | 8 | 864 | 4072487 | ||
BU855133.1 | - | 2 | 113 | |||
BX647566.1 | - | 7 | 816 | |||
CR536528.1 | - | 6 | 718 | |||
CR541880.1 | - | 6 | 718 | |||
EF036496.1 | - | 4 | 366 | |||
EF036497.1 | - | 3 | 297 | |||
FJ474908.1 | - | 8 | 885 | |||
M11560.1 | - | 8 | 885 | |||
M21190.1 | - | 3 | 304 | |||
X05236.1 | - | 7 | 864 |