Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004304.4 | NP_004295.2 | 27, 28 | 5025 | ||
GenBank mRNA | AB209477.4 | - | 27, 28 | 4562 | ||
EU788003.1 | - | 27, 28 | 4980 | |||
EU788004.1 | - | 27, 28 | 4980 | |||
EU788005.1 | - | 27, 28 | 4980 | |||
EU788006.1 | - | 27, 28 | 4980 | |||
EU788007.1 | - | 27, 28 | 4980 | |||
EU788008.1 | - | 27, 28 | 4980 | |||
EU788009.1 | - | 27, 28 | 4980 | |||
EU788010.1 | - | 27, 28 | 4980 | |||
FJ838786.1 | - | 27, 28 | 4980 | |||
FJ838787.1 | - | 27, 28 | 4980 | |||
GU128152.1 | - | 27, 28 | 4073 | |||
GU128153.1 | - | 27, 28 | 4073 | |||
GU128154.1 | - | 27, 28 | 4073 | |||
GU128155.1 | - | 27, 28 | 4073 | |||
GU128156.1 | - | 27, 28 | 4073 | |||
GU128157.1 | - | 27, 28 | 4073 | |||
LN864494.1 | - | 8, 9 | 1273 | |||
U62540.1 | - | 27, 28 | 4984 | |||
U66559.1 | - | 27, 28 | 4221 |