Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALOX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000698.4 | NP_000689.1 | 7 | 1044 | ||
NM_001256153.2 | NP_001243082.1 | 7 | 1044 | |||
NM_001256154.2 | NP_001243083.1 | 7 | 1044 | |||
NM_001320861.1 | NP_001307790.1 | 7 | 1044 | |||
NM_001320862.1 | NP_001307791.1 | 7 | 1082 | |||
XM_017016012.1 | XP_016871501.1 | 6 | 919 | |||
GenBank mRNA | AB208946.1 | - | 8 | 3315 | ||
AK315223.1 | - | 7 | 994 | |||
BC034464.1 | - | 7 | 983 | |||
BC040450.1 | - | 7 | 973 | |||
BC130332.1 | - | 7 | 997 | 40146363 | ||
BC132677.1 | - | 7 | 997 | 40146699 | ||
BC143985.1 | - | 7 | 997 | 9052503 | ||
HM592258.1 | - | 7 | 930 | |||
HM592259.1 | - | 7 | 930 | |||
HM592260.1 | - | 7 | 930 | |||
HM592261.1 | - | 7 | 930 | |||
J03571.1 | - | 7 | 964 | |||
J03600.1 | - | 7 | 974 | |||
JQ728550.1 | - | 6 | 850 |