Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257360.1 | NP_001244289.1 | 19 | 3150 | ||
NM_001257361.1 | NP_001244290.1 | 18 | 2433 | |||
NM_001308170.1 | NP_001295099.1 | 17 | 2553 | |||
NM_004037.7 | NP_004028.3 | 18 | 3017 | |||
NM_139156.3 | NP_631895.1 | 18 | 2797 | |||
NM_203404.1 | NP_981949.1 | 16 | 2435 | |||
GenBank mRNA | AK025706.1 | - | 18 | 2773 | ||
AK296394.1 | - | 18 | 2389 | |||
AK302939.1 | - | 18 | 2395 | |||
AK314494.1 | - | 19 | 3073 | |||
BC007711.2 | - | 18 | 2722 | 4101667 | ||
BC075844.1 | - | 18 | 2722 | 4130690 | ||
S47833.1 | - | 17 | 2408 | |||
U16270.1 | - | 13 | 1821 |