Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001636.3 | NP_001627.2 | 2 | 462 | ||
GenBank mRNA | AB209822.1 | - | 2 | 417 | ||
AF091236.1 | - | 1 | 183 | |||
AK312336.1 | - | 2 | 429 | |||
AY007135.1 | - | 2 | 414 | |||
BC007295.1 | - | 2 | 377 | 3349670 | ||
BC007850.2 | - | 2 | 416 | 4136545 | ||
BC008737.2 | - | 2 | 404 | 3342722 | ||
BC008935.2 | - | 2 | 409 | 2824067 | ||
BC013256.1 | - | 2 | 416 | |||
BC014775.1 | - | 2 | 417 | 3458777 | ||
BC031912.1 | - | 2 | 362 | 5141625 | ||
BC035469.1 | - | 2 | 416 | |||
CR457400.1 | - | 2 | 325 |