Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001154.3 | NP_001145.1 | 6 | 505 | ||
XM_017008141.1 | XP_016863630.1 | 6 | 505 | |||
GenBank mRNA | AK312644.1 | - | 6 | 505 | ||
AY577779.1 | - | 4 | 341 | |||
BC001429.2 | - | 5 | 410 | 3140878 | ||
BC004993.1 | - | 6 | 463 | 2906015 | ||
BC012804.2 | - | 6 | 457 | 2961097 | ||
BC012822.2 | - | 6 | 457 | 2961097 | ||
BC018671.1 | - | 6 | 497 | 4252480 | ||
BC032093.1 | - | 6 | 494 | |||
CR536522.1 | - | 4 | 340 | |||
CR541842.1 | - | 4 | 340 | |||
D00172.1 | - | 6 | 475 | |||
EU794590.1 | - | 6 | 505 | |||
J03745.1 | - | 6 | 499 | |||
M18366.1 | - | 5 | 352 | |||
M19384.1 | - | 6 | 490 | |||
M21731.1 | - | 6 | 482 | |||
X12454.1 | - | 6 | 468 |