Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001156.4 | NP_001147.1 | 10 | 1048 | ||
NM_001320879.1 | NP_001307808.1 | 9 | 993 | |||
NM_001320880.1 | NP_001307809.1 | 10 | 1059 | |||
NM_004034.3 | NP_004025.1 | 11 | 1114 | |||
XM_017016162.1 | XP_016871651.1 | 11 | 1150 | |||
XM_017016163.1 | XP_016871652.1 | 10 | 1084 | |||
GenBank mRNA | AB062429.1 | - | 10 | 1006 | ||
AK222552.1 | - | 10 | 1016 | |||
AK295319.1 | - | 10 | 925 | |||
AK300086.1 | - | 8 | 815 | |||
AK301681.1 | - | 10 | 802 | |||
AK307772.1 | - | 9 | 866 | |||
AK309141.1 | - | 10 | 810 | |||
AK312447.1 | - | 11 | 1087 | |||
AK313024.1 | - | 10 | 991 | |||
BC001322.1 | - | 3 | 299 | |||
BC002632.2 | - | 10 | 1008 | 3610541 | ||
BQ689410.1 | - | 2 | 112 | |||
BT007187.1 | - | 9 | 973 | |||
CR407686.1 | - | 9 | 973 | |||
J04543.1 | - | 10 | 1033 |