Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001156.4 | NP_001147.1 | 7 | 657 | ||
NM_001320879.1 | NP_001307808.1 | 6 | 602 | |||
NM_001320880.1 | NP_001307809.1 | 7 | 668 | |||
NM_004034.3 | NP_004025.1 | 8 | 723 | |||
XM_017016162.1 | XP_016871651.1 | 8 | 759 | |||
XM_017016163.1 | XP_016871652.1 | 7 | 693 | |||
GenBank mRNA | AB062429.1 | - | 7 | 615 | ||
AK222552.1 | - | 7 | 625 | |||
AK295319.1 | - | 7 | 534 | |||
AK300086.1 | - | 5 | 424 | |||
AK301681.1 | - | 7 | 411 | |||
AK307772.1 | - | 6 | 476 | |||
AK309141.1 | - | 7 | 419 | |||
AK312447.1 | - | 8 | 696 | |||
AK313024.1 | - | 7 | 600 | |||
BC002632.2 | - | 7 | 617 | 3610541 | ||
BT007187.1 | - | 6 | 582 | |||
BX361177.2 | - | 6 | 568 | |||
CR407686.1 | - | 6 | 582 | |||
J04543.1 | - | 7 | 642 |