Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001157.2 | NP_001148.1 | 15 | 1720 | ||
NM_001278407.1 | NP_001265336.1 | 17 | 1699 | |||
NM_001278408.1 | NP_001265337.1 | 17 | 2307 | |||
NM_001278409.1 | NP_001265338.1 | 17 | 2038 | |||
NM_145868.1 | NP_665875.1 | 16 | 1769 | |||
NM_145869.1 | NP_665876.1 | 17 | 1965 | |||
XM_005269741.4 | XP_005269798.1 | 13 | 1797 | |||
XM_011539736.2 | XP_011538038.1 | 17 | 2248 | |||
GenBank mRNA | AB209770.1 | - | 16 | 1714 | ||
AJ278463.1 | - | 15 | 1719 | |||
AJ278464.1 | - | 16 | 1768 | |||
AJ278465.1 | - | 17 | 1964 | |||
AK000567.1 | - | 15 | 1719 | |||
AK056107.1 | - | 17 | 1699 | |||
AK126287.1 | - | 17 | 2307 | |||
AK222918.1 | - | 15 | 1682 | |||
AK298362.1 | - | 16 | 1530 | |||
AK300509.1 | - | 13 | 1832 | |||
AK301047.1 | - | 17 | 2023 | |||
BC007564.2 | - | 16 | 1555 | 2989058 | ||
BT019934.1 | - | 14 | 1482 | |||
CR450323.1 | - | 14 | 1482 | |||
L19605.1 | - | 15 | 1660 |