Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003954.2 | NP_001003954.1 | 10 | 901 | ||
NM_004306.3 | NP_004297.2 | 9 | 778 | |||
GenBank mRNA | AJ306450.1 | - | 10 | 877 | ||
AK223374.1 | - | 9 | 777 | |||
AK290281.1 | - | 10 | 887 | |||
AW854063.1 | - | 3 | 596 | |||
BC125158.1 | - | 9 | 748 | 40120955 | ||
CR541838.1 | - | 8 | 686 | |||
Z11502.1 | - | 9 | 765 |