Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163.3 | NP_001154.2 | 3 | 1555 | ||
XM_005251968.3 | XP_005252025.1 | 3 | 1586 | |||
XM_011518617.2 | XP_011516919.1 | 3 | 1510 | |||
XM_017014670.1 | XP_016870159.1 | 3 | 1799 | |||
GenBank mRNA | AF047347.1 | - | 3 | 1518 |