Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOBEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304566.1 | NP_001291495.1 | 3 | 430 | ||
NM_001644.4 | NP_001635.2 | 3 | 437 | |||
NM_005889.3 | NP_005880.2 | 2 | 409 | |||
GenBank mRNA | AI380263.1 | - | 3 | 460 | ||
AI862416.1 | - | 3 | 468 | |||
BC096158.1 | - | 3 | 427 | |||
BC096160.1 | - | 2 | 399 | |||
BC096161.1 | - | 2 | 399 | |||
BC101404.1 | - | 2 | 396 | |||
BC101405.1 | - | 2 | 395 | |||
BC101972.1 | - | 3 | 424 | |||
BC101973.1 | - | 3 | 427 | |||
BC144708.1 | - | 2 | 409 | |||
L25877.1 | - | 3 | 428 | |||
L26234.1 | - | 3 | 430 | |||
U72891.1 | - | 3 | 428 | |||
U78720.1 | - | 1 | 362 |