Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169.2 | NP_001160.2 | 5 | 804 | ||
XM_011545822.2 | XP_011544124.1 | 5 | 847 | |||
XM_011545823.2 | XP_011544125.1 | 3 | 512 | |||
GenBank mRNA | AB013456.1 | - | 5 | 806 | ||
AF067797.1 | - | 5 | 807 | |||
AK222964.1 | - | 5 | 769 | |||
AK222975.1 | - | 5 | 801 | |||
BC040630.1 | - | 5 | 772 | 5759105 | ||
BG221545.1 | - | 1 | 124 |