Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHROOM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320663.1 | NP_001307592.1 | 2, 3 | 902 | ||
NM_001320664.1 | NP_001307593.1 | 2, 3 | 902 | |||
NM_001649.3 | NP_001640.1 | 6, 7 | 3667 | |||
XM_005274500.4 | XP_005274557.2 | 6, 7 | 3717 | |||
XM_011545506.1 | XP_011543808.1 | 2, 3 | 779 | |||
XM_017029492.1 | XP_016884981.1 | 2, 3 | 779 | |||
XM_017029493.1 | XP_016884982.1 | 2, 3 | 1272 | |||
XM_017029494.1 | XP_016884983.1 | 2, 3 | 1272 | |||
GenBank mRNA | AK096596.1 | - | 2, 3 | 1233 | ||
AK299907.1 | - | 2, 3 | 902 | |||
BC140866.1 | - | 6, 7 | 3628 | 9021734 | ||
CR749271.1 | - | 2, 3 | 841 | |||
X83543.1 | - | 6, 7 | 3667 |