Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001651.3 | NP_001642.1 | 1, 2 | 887 | ||
XM_005268838.2 | XP_005268895.1 | 1, 2 | 528 | |||
GenBank mRNA | BC032946.1 | - | 1, 2 | 437 | 5269384 | |
CR541989.1 | - | 1, 2 | 365 | |||
CR542022.1 | - | 1, 2 | 365 |