Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320635.1 | NP_001307564.1 | 1 | 412 | ||
NM_020980.4 | NP_066190.2 | 1 | 412 | |||
GenBank mRNA | AB008775.1 | - | 1 | 258 | ||
AF016495.1 | - | 1 | 270 | |||
AK313214.1 | - | 1 | 337 | |||
BC026258.1 | - | 1 | 227 | 4734559 | ||
CR542027.1 | - | 1 | 42 | |||
DC410284.1 | - | 1 | 412 |