Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320635.1 | NP_001307564.1 | 4 | 825 | ||
NM_001320636.1 | NP_001307565.1 | 4 | 457 | |||
NM_020980.4 | NP_066190.2 | 4 | 825 | |||
GenBank mRNA | AB008775.1 | - | 4 | 671 | ||
AF016495.1 | - | 4 | 683 | |||
AK300055.1 | - | 4 | 430 | |||
AK313214.1 | - | 4 | 750 | |||
BC026258.1 | - | 4 | 640 | 4734559 | ||
CB528930.1 | - | 3 | 676 | |||
CR542027.1 | - | 4 | 455 | |||
DC296768.1 | - | 4 | 461 |