Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000044.3 | NP_000035.2 | 4 | 3211 | ||
NM_001011645.2 | NP_001011645.1 | 4 | 662 | |||
GenBank mRNA | AF162704.1 | - | 6 | 2057 | ||
BC132975.1 | - | 4 | 2541 | 40146997 | ||
GU208210.1 | - | 4 | 2114 | |||
HQ692823.1 | - | 4 | 500 | |||
KF720403.1 | - | 1 | 122 | |||
KF720404.1 | - | 1 | 122 | |||
KF720405.1 | - | 1 | 122 | |||
KJ534771.1 | - | 4 | 2093 | |||
L29496.1 | - | 4 | 1538 | |||
M20132.1 | - | 4 | 2455 | |||
M20260.1 | - | 4 | 692 | |||
M21748.1 | - | 4 | 2249 | |||
M23263.1 | - | 4 | 2621 | |||
M34233.1 | - | 6 | 2162 | |||
M73069.1 | - | 5 | 2120 |