Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256196.1 | NP_001243125.1 | 5 | 549 | ||
NM_001256197.1 | NP_001243126.1 | 5 | 549 | |||
NM_001654.4 | NP_001645.1 | 5 | 549 | |||
XM_006724529.2 | XP_006724592.1 | 5 | 470 | |||
XM_011543906.2 | XP_011542208.1 | 5 | 1860 | |||
XM_011543908.2 | XP_011542210.1 | 5 | 1859 | |||
GenBank mRNA | AB158254.1 | - | 4 | 346 | ||
AB208831.1 | - | 5 | 446 | |||
AK130043.1 | - | 5 | 442 | |||
AK297453.1 | - | 5 | 442 | |||
AK300975.1 | - | 5 | 446 | |||
AK307747.1 | - | 5 | 442 | |||
BC002466.2 | - | 5 | 429 | 3347509 | ||
BC007514.2 | - | 5 | 427 | 2966604 | ||
BT019864.1 | - | 4 | 338 | |||
DA264472.1 | - | 5 | 471 | |||
DC362427.1 | - | 5 | 549 | |||
KU177914.1 | - | 4 | 338 | |||
KU177915.1 | - | 4 | 338 | |||
X04790.1 | - | 5 | 532 |