Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256196.1 | NP_001243125.1 | 9 | 963 | ||
NM_001654.4 | NP_001645.1 | 9 | 954 | |||
XM_006724529.2 | XP_006724592.1 | 9 | 890 | |||
XM_011543906.2 | XP_011542208.1 | 9 | 2280 | |||
XM_011543908.2 | XP_011542210.1 | 9 | 2264 | |||
XM_011543909.2 | XP_011542211.1 | 4 | 255 | |||
GenBank mRNA | AB208831.1 | - | 8 | 2139 | ||
AK130043.1 | - | 9 | 847 | |||
AK297453.1 | - | 7 | 677 | |||
AK300975.1 | - | 9 | 810 | |||
AK307747.1 | - | 9 | 848 | |||
BC002466.2 | - | 9 | 834 | 3347509 | ||
BC007514.2 | - | 9 | 841 | 2966604 | ||
BT019864.1 | - | 8 | 743 | |||
KU177914.1 | - | 8 | 743 | |||
KU177915.1 | - | 8 | 758 | |||
X04790.1 | - | 9 | 937 |