Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001662.3 | NP_001653.1 | 4 | 418 | ||
GenBank mRNA | AF493884.1 | - | 4 | 271 | ||
AK297026.1 | - | 4 | 323 | |||
AK307297.1 | - | 3 | 863 | |||
AK308548.1 | - | 3 | 1071 | |||
AK312107.1 | - | 4 | 323 | |||
BC003043.1 | - | 4 | 308 | 2822973 | ||
BC033104.1 | - | 4 | 365 | 4862014 | ||
BT007087.1 | - | 4 | 271 | |||
CN259625.1 | - | 4 | 435 | |||
M57567.1 | - | 4 | 325 |