Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313941.1 | NP_001300870.1 | 4 | 606 | ||
NM_001313943.1 | NP_001300872.1 | 3 | 511 | |||
NM_001313944.1 | NP_001300873.1 | 3 | 451 | |||
NM_001313945.1 | NP_001300874.1 | 3 | 388 | |||
NM_001664.3 | NP_001655.1 | 3 | 511 | |||
GenBank mRNA | AF498970.1 | - | 2 | 231 | ||
AK130066.1 | - | 3 | 507 | |||
AK130808.1 | - | 3 | 387 | |||
AK222556.1 | - | 3 | 403 | |||
AK296654.1 | - | 3 | 327 | |||
AK307220.1 | - | 3 | 286 | |||
BC001360.2 | - | 3 | 343 | 3051460 | ||
BC005976.1 | - | 3 | 348 | 4102976 | ||
BF966305.1 | - | 4 | 481 | |||
BQ721697.1 | - | 3 | 384 | |||
BT019870.1 | - | 2 | 231 | |||
BX647063.1 | - | 3 | 407 | |||
L09159.1 | - | 2 | 220 | |||
L25080.1 | - | 3 | 382 | |||
X05026.1 | - | 3 | 389 |