Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254738.1 | NP_001241667.1 | 2, 3 | 516 | ||
NM_005168.4 | NP_005159.1 | 3, 4 | 423 | |||
GenBank mRNA | AF498969.1 | - | 2, 3 | 238 | ||
AK299731.1 | - | 2, 3 | 223 | |||
AK313224.1 | - | 2, 3 | 490 | |||
AL702945.1 | - | 2, 3 | 489 | |||
BC012513.1 | - | 3, 4 | 378 | 4453777 | ||
BQ440721.1 | - | 2, 3 | 492 | |||
BT006769.1 | - | 2, 3 | 238 | |||
CR542038.1 | - | 2, 3 | 193 | |||
CR542070.1 | - | 2, 3 | 193 | |||
DC367501.1 | - | 3, 4 | 424 | |||
S82240.1 | - | 2, 3 | 217 | |||
X95282.1 | - | 2, 3 | 238 | |||
X97758.1 | - | 2, 3 | 460 |