Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164741.1 | NP_001158213.1 | 10 | 1327 | ||
NM_001666.4 | NP_001657.3 | 9 | 1207 | |||
GenBank mRNA | AK294562.1 | - | 9 | 1132 | ||
AY183123.1 | - | 9 | 1191 | |||
BC052303.1 | - | 10 | 1327 | 6379390 | ||
D50921.1 | - | 9 | 1138 | |||
X78817.1 | - | 9 | 1191 |