Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164741.1 | NP_001158213.1 | 22 | 2770 | ||
NM_001666.4 | NP_001657.3 | 21 | 2650 | |||
GenBank mRNA | AK294562.1 | - | 21 | 2575 | ||
AY183123.1 | - | 18 | 2216 | |||
BC023626.2 | - | 7 | 824 | |||
BC052303.1 | - | 22 | 2770 | 6379390 | ||
D50921.1 | - | 21 | 2581 | |||
X78817.1 | - | 21 | 2634 |