Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287242.1 | NP_001274171.1 | 10 | 1515 | ||
NM_006125.2 | NP_006116.2 | 10 | 2738 | |||
NM_013423.2 | NP_038267.1 | 10 | 1567 | |||
NM_013427.2 | NP_038286.2 | 10 | 2738 | |||
NR_109776.1 | 11 | 2834 | ||||
GenBank mRNA | AB208792.1 | - | 10 | 2120 | ||
AF012272.2 | - | 10 | 2739 | |||
AF022212.2 | - | 11 | 2835 | |||
AF117067.1 | - | 10 | 2739 | |||
AF177663.1 | - | 10 | 1567 | |||
AF177665.1 | - | 10 | 1373 | |||
AK293078.1 | - | 10 | 2169 | |||
AK297713.1 | - | 10 | 1412 | |||
AK297747.1 | - | 4 | 524 | |||
AK303444.1 | - | 10 | 1428 | |||
BC150635.1 | - | 10 | 1897 | 9057005 |