Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARNT SIRNA Assays ›
Gene Symbol:
ARNT
Gene Name:
aryl hydrocarbon receptor nuclear translocator
Gene Aliases:
HIF-1-beta, HIF-1beta, HIF1-beta, HIF1B, HIF1BETA, TANGO, bHLHe2
Chromosome Location:
Chr.1: 150809705 - 150876768 on Build GRCh38
UniGene ID:
Hs.632446
Species:
Human
siRNA ID
HSS100700
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARNT
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001197325.1
NP_001184254.1
6
783
NM_001286035.1
NP_001272964.1
8
884
NM_001286036.1
NP_001272965.1
7
828
NM_001668.3
NP_001659.1
7
828
NM_178427.2
NP_848514.1
6
783
XM_005245151.1
XP_005245208.1
7
770
XM_005245153.1
XP_005245210.1
7
770
XM_005245154.3
XP_005245211.1
8
798
XM_005245157.1
XP_005245214.1
7
770
XM_011509542.2
XP_011507844.1
7
831
XM_011509543.2
XP_011507845.1
8
826
XM_011509544.2
XP_011507846.1
7
795
XM_011509545.2
XP_011507847.1
6
786
XM_011509546.1
XP_011507848.1
6
546
XM_011509547.2
XP_011507849.1
6
786
XM_017001288.1
XP_016856777.1
8
673
XM_017001289.1
XP_016856778.1
8
673
XM_017001290.1
XP_016856779.1
7
627
XM_017001291.1
XP_016856780.1
7
629
XM_017001292.1
XP_016856781.1
8
674
XM_017001293.1
XP_016856782.1
7
631
XM_017001294.1
XP_016856783.1
7
770
XM_017001295.1
XP_016856784.1
7
770
XM_017001296.1
XP_016856785.1
6
734
GenBank mRNA
AB209877.1
-
7
639
AF001307.1
-
7
682
AK223459.1
-
692
AK290177.1
-
7
767
AK291705.1
-
6
625
AK293027.1
-
7
828
AK304772.1
-
6
617
AK313780.1
-
7
769
AK315812.1
-
6
622
AL834279.1
-
8
823
BC041121.1
-
7
642
BC060838.1
-
7
763
5300334
M69238.1
-
7
683
Back To Top
More Information
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
embryonic placenta development
positive regulation of endothelial cell proliferation
transcription, DNA-templated
positive regulation of vascular endothelial growth factor production
cell differentiation
intracellular receptor signaling pathway
positive regulation of vascular endothelial growth factor receptor signaling pathway
positive regulation of protein sumoylation
mRNA transcription from RNA polymerase II promoter
regulation of transcription from RNA polymerase II promoter in response to oxidative stress
positive regulation of erythrocyte differentiation
positive regulation of glycolytic process
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of hormone biosynthetic process
regulation of transcription from RNA polymerase II promoter in response to hypoxia
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription coactivator activity
aryl hydrocarbon receptor activity
protein binding
transcription factor binding
aryl hydrocarbon receptor binding
enhancer binding
sequence-specific DNA binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0