Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127505.2 | NP_001120977.1 | 7, 8 | 679 | ||
NM_004315.5 | NP_004306.3 | 7, 8 | 745 | |||
NM_177924.4 | NP_808592.2 | 7, 8 | 812 | |||
XM_005273504.3 | XP_005273561.1 | 7, 8 | 573 | |||
GenBank mRNA | AK025732.1 | - | 8, 9 | 715 | ||
AK098299.1 | - | 7, 8 | 460 | |||
AK222780.1 | - | 8, 9 | 548 | |||
AK289481.1 | - | 8, 9 | 713 | |||
AY305384.1 | - | 7, 8 | 586 | |||
BC016481.1 | - | 8, 9 | 535 | 3923451 | ||
BC016828.1 | - | 7, 8 | 666 | 4109317 | ||
BC035453.1 | - | 2, 3 | 110 | |||
U47674.1 | - | 7, 8 | 516 | |||
U70063.1 | - | 8, 9 | 517 |