Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286164.1 | NP_001273093.1 | 12 | 2261 | ||
NM_004319.2 | NP_004310.1 | 12 | 2261 | |||
NM_207108.2 | NP_996991.1 | 12 | 2261 | |||
XM_017001340.1 | XP_016856829.1 | 12 | 2235 | |||
XM_017001341.1 | XP_016856830.1 | 12 | 2235 | |||
GenBank mRNA | AB006627.1 | - | 12 | 2059 | ||
AK295126.1 | - | 12 | 2213 | |||
AK307820.1 | - | 12 | 2171 | |||
AK309094.1 | - | 12 | 2330 | |||
BC142697.1 | - | 12 | 2040 | 40148746 | ||
BC146795.1 | - | 12 | 2059 | 9007266 |