Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005171.4 | NP_005162.1 | 2 | 378 | ||
XM_011538386.2 | XP_011536688.1 | 2 | 2435 | |||
XM_011538387.2 | XP_011536689.1 | 2 | 108 | |||
XM_011538388.2 | XP_011536690.1 | 2 | 298 | |||
XM_017019331.1 | XP_016874820.1 | 2 | 148 | |||
XM_017019332.1 | XP_016874821.1 | 2 | 242 | |||
XM_017019333.1 | XP_016874822.1 | 2 | 128 | |||
XM_017019334.1 | XP_016874823.1 | 2 | 154 | |||
XM_017019335.1 | XP_016874824.1 | 2 | 141 | |||
XM_017019336.1 | XP_016874825.1 | 2 | 136 | |||
GenBank mRNA | AK299240.1 | - | 3 | 229 | ||
AK313406.1 | - | 2 | 283 | |||
BC029619.1 | - | 2 | 92 | 4102231 | ||
BG502726.1 | - | 2 | 111 | |||
BG545632.1 | - | 2 | 306 | |||
BI712653.1 | - | 2 | 267 | |||
DA468394.1 | - | 2 | 378 |